[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]methanamine ≥95%

Supplier: Matrix Scientific

044277-500MG 044277-1G 044277-5G
101865-052EA 747.69 USD
101865-052 101865-054 101865-056
[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]methanamine ≥95%
[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]methanamine

Hazard: Irritant

Formula: C₁₁H₁₁N₂ClS
Melting Pt: 117-119°C
MDL Number: MFCD03791207
CAS Number: 886360-58-3

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR