Dimethyl-2-[2,6-dinitro-4-(trifluoromethyl)phenyl]malonate ≥95%

Supplier: Matrix Scientific

044275-500MG 044275-1G 044275-5G
101865-042EA 278.46 USD
101865-042 101865-044 101865-046
Dimethyl-2-[2,6-dinitro-4-(trifluoromethyl)phenyl]malonate ≥95%
Dimethyl-2-[2,6-dinitro-4-(trifluoromethyl)phenyl]malonate

Hazard: Irritant

Formula: C₁₂H₉F₃N₂O₈
Melting Pt: 101…103 °C
MDL Number: MFCD03787071
CAS Number: 865658-18-0

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR