2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥95%
Supplier: Matrix Scientific
044273-500MG
044273-1G
101865-038EA
921.54
USD
101865-038
101865-040
2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥95%
2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole
Hazard: Irritant
Formula:
C₃H₃F₃N₄S Melting Pt: 129…132 °C |
MDL Number:
MFCD07786303 CAS Number: 299440-05-4 |
Specification Test Results
Melting point | 129-132°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...