5-Bromobenzo[b]thiophene-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
044272-500MG
044272-1G
044272-5G
101865-032EA
163.08
USD
101865-032
101865-034
101865-036
5-Bromobenzo[b]thiophene-2-carboxylic acid ≥95%
5-Bromobenzo[b]thiophene-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅BrO₂S MW: 257.1000 g/mol Melting Pt: 230…234 °C |
MDL Number:
MFCD01929338 CAS Number: 7312-10-9 |
Specification Test Results
Melting point | 230-234°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...