3-(Dimethylamino)-2-(2-pyridinyl)acrylonitrile ≥95%

Supplier: Matrix Scientific

044270-500MG 044270-1G 044270-5G
101865-020EA 215.38 USD
101865-020 101865-022 101865-024
3-(Dimethylamino)-2-(2-pyridinyl)acrylonitrile ≥95%
3-(Dimethylamino)-2-(2-pyridinyl)acrylonitrile

Hazard: Irritant

Formula: C₁₀H₁₁N₃
Melting Pt: 70…72 °C
MDL Number: MFCD01570039
CAS Number: 493038-85-0

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR