2-(3-Nitrophenyl)quinoxaline ≥95%

Supplier: Matrix Scientific

044267-500MG 044267-1G
101865-010EA 278.46 USD
101865-010 101865-012
2-(3-Nitrophenyl)quinoxaline ≥95%
2-(3-Nitrophenyl)quinoxaline

Hazard: Irritant

Formula: C₁₄H₉N₃O₂
Melting Pt: 184-185°C
MDL Number: MFCD00430477
CAS Number: 5021-44-3

Order Now

Specification Test Results

Melting point 184-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR