Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044266-500MG 044266-1G 044266-5G
101865-004EA 747.69 USD
101865-004 101865-006 101865-008
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate ≥95%
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₃F₃N₂O₄
MW: 318.25 g/mol
Melting Pt: 231…233 °C
MDL Number: MFCD03791206
CAS Number: 886360-57-2

Order Now

Specification Test Results

Melting point 231-233°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR