3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)-3-(trifluoroacetamido)propanoic acid

044265-500MG 044265-1G 044265-5G
101864-998EA 278.46 USD
101864-998 101865-000 101865-002
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid

Hazard: Irritant

Formula: C₁₁H₉ClF₃NO₃
MW: 295.65 g/mol
Melting Pt: 162…165 °C
MDL Number: MFCD03791205
CAS Number: 117291-25-5

Order Now

Specification Test Results

Melting point 162-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR