3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%

Supplier: Matrix Scientific

044264-500MG 044264-1G 044264-5G
101864-992EA 556.92 USD
101864-992 101864-994 101864-996
3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%
3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₂F₃NO₅
Melting Pt: 170…173 °C
MDL Number: MFCD03791204
CAS Number: 886360-56-1

Order Now

Specification Test Results

Melting point 170-173°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR