7-Nitro-2H-1,4-benzoxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

044258-500MG 044258-1G 044258-5G
101864-976EA 24.62 USD
101864-976 101864-978 101864-980
7-Nitro-2H-1,4-benzoxazin-3(4H)-one ≥95%
7-Nitro-2H-1,4-benzoxazin-3(4H)-one

Hazard: Irritant

Formula: C₈H₆N₂O₄
Melting Pt: 232…234 °C
MDL Number: MFCD00090355
CAS Number: 81721-86-0

Order Now

Specification Test Results

Melting point 232-234°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR