5-(4-Morpholinyl)-2-nitroaniline ≥95%

Supplier: Matrix Scientific

044257-500MG 044257-1G 044257-5G
101864-970EA 129.23 USD
101864-970 101864-972 101864-974
5-(4-Morpholinyl)-2-nitroaniline ≥95%
5-(4-Morpholinyl)-2-nitroaniline

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₃
Melting Pt: 182…185 °C
MDL Number: MFCD00511844
CAS Number: 54998-00-4

Order Now

Specification Test Results

Melting point 182-185°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR