[6-(4-Bromophenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol ≥95%

Supplier: Matrix Scientific

044256-500MG 044256-1G 044256-5G
101864-964EA 556.92 USD
101864-964 101864-966 101864-968
[6-(4-Bromophenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol ≥95%
[6-(4-Bromophenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol

Hazard: Irritant

Formula: C₁₂H₉BrN₂OS
Melting Pt: 197…199 °C
MDL Number: MFCD03425764
CAS Number: 439095-24-6

Order Now

Specification Test Results

Melting point 197-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR