2-Oxo-2-phenylethyl-2-aminobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
044254-500MG
044254-1G
101864-954EA
181.54
USD
101864-954
101864-956
2-Oxo-2-phenylethyl-2-aminobenzenecarboxylate ≥95%
2-Oxo-2-phenylethyl-2-aminobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₃NO₃ Melting Pt: 119…123 °C |
MDL Number:
MFCD00736387 CAS Number: 130627-10-0 |
Specification Test Results
Melting point | 119-123°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...