[6-(4-Methylphenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol ≥95%
Supplier: Matrix Scientific
044252-500MG
044252-1G
044252-5G
101864-948EA
556.92
USD
101864-948
101864-950
101864-952
[6-(4-Methylphenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol ≥95%
[6-(4-Methylphenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol
Hazard: Irritant
Formula:
C₁₃H₁₂N₂OS Melting Pt: 192…194 °C |
MDL Number:
MFCD03425761 CAS Number: 82588-60-1 |
Specification Test Results
Melting point | 192-194°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...