(4-Imidazol-1-ylphenyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(1H-Imidazol-1-yl)benzyl alcohol, (4-Imidazol-1-yl-phenyl)methanol

044251-500MG 044251-1G 044251-5G
101864-942EA 278.46 USD
101864-942 101864-944 101864-946
(4-Imidazol-1-ylphenyl)methanol ≥95%
(4-Imidazol-1-ylphenyl)methanol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2000 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD02682059
CAS Number: 86718-08-3

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR