4-Amino-2-(hydroxymethyl)benzenol ≥95%

Supplier: Matrix Scientific

044246-500MG 044246-1G 044246-5G
101864-920EA 556.92 USD
101864-920 101864-922 101864-924
4-Amino-2-(hydroxymethyl)benzenol ≥95%
4-Amino-2-(hydroxymethyl)benzenol

Hazard: Irritant

Formula: C₇H₉NO₂
Melting Pt: 147…149 °C
MDL Number: MFCD03425760
CAS Number: 104333-09-7

Order Now

Specification Test Results

Melting point 147-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR