4-Imidazol-1-ylbenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(1-Imidazolyl)benzaldehyde, 1-(4-Formylphenyl)imidazole, 4-(1H-Imidazol-1-yl)benzaldehyde

044245-500MG 044245-1G 044245-5G
101864-914EA 20 USD
101864-914 101864-916 101864-918
4-Imidazol-1-ylbenzaldehyde ≥95%
4-Imidazol-1-ylbenzaldehyde

Hazard: Irritant

Formula: C₁₀H₈N₂O
MW: 172.1833 g/mol
Melting Pt: 148…150 °C
MDL Number: MFCD00209977
CAS Number: 10040-98-9

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR