Methyl-4-{2-amino-4-[bis(acetyloxy)methyl]phenoxy}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044243-500MG 044243-1G 044243-5G
101864-902EA 747.69 USD
101864-902 101864-904 101864-906
Methyl-4-{2-amino-4-[bis(acetyloxy)methyl]phenoxy}benzenecarboxylate ≥95%
Methyl-4-{2-amino-4-[bis(acetyloxy)methyl]phenoxy}benzenecarboxylate

Hazard: Irritant

Formula: C₁₉H₁₉NO₇
Melting Pt: 105…107 °C
MDL Number: MFCD03425759
CAS Number: 451485-65-7

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR