2-(4-Methylpiperazino)-3-pyridinamine ≥95%

Supplier: Matrix Scientific

044239-500MG 044239-1G 044239-5G
101864-884EA 556.92 USD
101864-884 101864-886 101864-888
2-(4-Methylpiperazino)-3-pyridinamine ≥95%
2-(4-Methylpiperazino)-3-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₆N₄
Melting Pt: 112…114 °C
MDL Number: MFCD01704018
CAS Number: 5028-17-1

Order Now

Specification Test Results

Melting point 112-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR