2-(Hydroxymethyl)-4-nitrobenzenol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitrosalicyl alcohol, 2-(Hydroxymethyl)-4-nitrophenol, 2-Hydroxy-5-nitrobenzyl alcohol

044238-500MG 044238-1G 044238-5G
101864-878EA 56.92 USD
101864-878 101864-880 101864-882
2-(Hydroxymethyl)-4-nitrobenzenol ≥95%
2-(Hydroxymethyl)-4-nitrobenzenol

Hazard: Irritant

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD01321361
CAS Number: 39224-61-8

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR