Methyl-4-(4-formyl-2-nitrophenoxy)-3-methoxybenzenecarboxylate ≥95%

Supplier: Matrix Scientific

044237-500MG 044237-1G 044237-5G
101864-872EA 278.46 USD
101864-872 101864-874 101864-876
Methyl-4-(4-formyl-2-nitrophenoxy)-3-methoxybenzenecarboxylate ≥95%
Methyl-4-(4-formyl-2-nitrophenoxy)-3-methoxybenzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₃NO₇
Melting Pt: 117…118 °C
MDL Number: MFCD03425757
CAS Number: 439095-22-4

Order Now

Specification Test Results

Melting point 117-118°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR