7-(Diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 7-(Diethylamino)-4-(trifluoromethyl)coumarin-3-carboxylic acid

044233-500MG 044233-1G
101864-856EA 747.69 USD
101864-856 101864-858
7-(Diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid ≥95%
7-(Diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₄F₃NO₄
MW: 329.28 g/mol
Melting Pt: 180…182 °C
MDL Number: MFCD03425755
CAS Number: 333303-14-3

Order Now

Specification Test Results

Melting point 180-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR