1-[4-(Diethylamino)-2-hydroxyphenyl]-2,2,2-trifluoro-1-ethanone ≥95%
Supplier: Matrix Scientific
044230-500MG
044230-1G
101864-844EA
278.46
USD
101864-844
101864-846
1-[4-(Diethylamino)-2-hydroxyphenyl]-2,2,2-trifluoro-1-ethanone ≥95%
1-[4-(Diethylamino)-2-hydroxyphenyl]-2,2,2-trifluoro-1-ethanone
Hazard: Irritant
Formula:
C₁₂H₁₄F₃NO₂ Melting Pt: 51…52 °C |
MDL Number:
MFCD08056623 CAS Number: 333303-11-0 |
Specification Test Results
Melting point | 51-52°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...