N-Methyl-2-phenylacetamide ≥95%

Supplier: Matrix Scientific

044226-500MG 044226-1G 044226-5G
101864-824EA 383.08 USD
101864-824 101864-826 101864-828
N-Methyl-2-phenylacetamide ≥95%
N-Methyl-2-phenylacetamide

Hazard: Irritant

Formula: C₉H₁₁NO
Melting Pt: 58…60 °C
MDL Number: MFCD00051354
CAS Number: 6830-82-6

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR