Methyl-1-(phenylsulfonyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044224-500MG 044224-1G 044224-5G
101864-812EA 278.46 USD
101864-812 101864-814 101864-816
Methyl-1-(phenylsulfonyl)-4-piperidinecarboxylate ≥95%
Methyl-1-(phenylsulfonyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₇NO₄S
Melting Pt: 83…85 °C
MDL Number: MFCD01045646
CAS Number: 311794-15-7

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR