Methyl-1-[(4-fluorophenyl)sulfonyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044223-500MG 044223-1G 044223-5G
101864-806EA 278.46 USD
101864-806 101864-808 101864-810
Methyl-1-[(4-fluorophenyl)sulfonyl]-4-piperidinecarboxylate ≥95%
Methyl-1-[(4-fluorophenyl)sulfonyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆FNO₄S
Melting Pt: 90…91 °C
MDL Number: MFCD02736084
CAS Number: 349624-64-2

Order Now

Specification Test Results

Melting point 90-91°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR