Methyl-1-[(4-methylphenyl)sulfonyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044221-500MG 044221-1G 044221-5G
101864-794EA 278.46 USD
101864-794 101864-796 101864-798
Methyl-1-[(4-methylphenyl)sulfonyl]-4-piperidinecarboxylate ≥95%
Methyl-1-[(4-methylphenyl)sulfonyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₄S
Melting Pt: 125…127 °C
MDL Number: MFCD01045650
CAS Number: 311794-12-4

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR