Methyl-2-acetyl-3-(dimethylamino)acrylate ≥95%

Supplier: Matrix Scientific

044219-1G 044219-5G
101864-784EA 106.15 USD
101864-784 101864-786
Methyl-2-acetyl-3-(dimethylamino)acrylate ≥95%
Methyl-2-acetyl-3-(dimethylamino)acrylate

Hazard: Irritant

Formula: C₈H₁₃NO₃
Melting Pt: 66…67 °C
MDL Number: MFCD08689663
CAS Number: 203186-56-5

Order Now

Specification Test Results

Melting point 66-67°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR