1-(Phenylsulfonyl)-4-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(Phenylsulphonyl)piperidine-4-carboxylic acid
044218-500MG
044218-1G
044218-5G
101864-776EA
375.38
USD
101864-776
101864-778
101864-780
1-(Phenylsulfonyl)-4-piperidinecarboxylic acid ≥95%
1-(Phenylsulphonyl)-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₅NO₄S MW: 269.3168 g/mol Melting Pt: 157…159.5 °C |
MDL Number:
MFCD00723116 CAS Number: 122891-92-3 |
Specification Test Results
Melting point | 157-159.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...