Methyl-4-methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-methyl-2-(methylthio)pyrimidine-5-carboxylate

044217-500MG 044217-1G 044217-5G
101864-770EA 180 USD
101864-770 101864-772 101864-774
Methyl-4-methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylate ≥95%
Methyl-4-methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂S
MW: 198.25 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD04125151
CAS Number: 166392-24-1

Order Now

Specification Test Results

Melting point 59-61°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR