Ethyl-3,6-dichloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

044216-500MG 044216-1G 044216-5G
101864-764EA 375.38 USD
101864-764 101864-766 101864-768
Ethyl-3,6-dichloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 3,6-dichloroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈Cl₂N₂O₂
MW: 259.887 g/mol
Melting Pt: 148-150°C
MDL Number: MFCD02186226
CAS Number: 478040-91-4

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR