Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

044213-500MG 044213-1G 044213-5G
101864-752EA 278.46 USD
101864-752 101864-754 101864-756
Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂
Melting Pt: 112-114°C
MDL Number: MFCD02186220
CAS Number: 67625-38-1

Order Now

Specification Test Results

Melting point 112-114°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR