Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate ≥95%

Supplier: Matrix Scientific

044211-500MG 044211-1G
101864-742EA 1109.23 USD
101864-742 101864-744
Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate ≥95%
Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate

Hazard: Irritant

Formula: C₉H₉N₃O₂
Melting Pt: 170…172 °C
MDL Number: MFCD02186164
CAS Number: 64951-06-0

Order Now

Specification Test Results

Melting point 170-172°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR