Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 7-methylimidazo[1,2-a]pyridine-2-carboxylate

044210-500MG 044210-1G
101864-738EA 556.92 USD
101864-738 101864-740
Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 7-methylimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MW: 204.2300 g/mol
Melting Pt: 126-129°C
MDL Number: MFCD02187526
CAS Number: 70705-33-8

Order Now

Specification Test Results

Melting point 126-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR