6-{[2-(4-Chlorobenzoyl)hydrazino]carbonyl}-3-cyclohexene-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

044205-500MG 044205-1G 044205-5G
101864-710EA 278.46 USD
101864-710 101864-712 101864-714
6-{[2-(4-Chlorobenzoyl)hydrazino]carbonyl}-3-cyclohexene-1-carboxylic acid ≥95%
6-{[2-(4-Chlorobenzoyl)hydrazino]carbonyl}-3-cyclohexene-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O₄
Melting Pt: 180…182 °C
MDL Number: MFCD02186201
CAS Number: 451485-61-3

Order Now

Specification Test Results

Melting point 180-182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR