3-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1H-pyrazole ≥95%

Supplier: Matrix Scientific

044204-500MG 044204-1G 044204-5G
101864-704EA 375.38 USD
101864-704 101864-706 101864-708
3-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1H-pyrazole ≥95%
3-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1H-pyrazole

Hazard: Irritant

Formula: C₁₆H₁₂ClFN₂O
Melting Pt: 125…127 °C
MDL Number: MFCD03425750
CAS Number: 451485-60-2

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR