1,2-Dihydronaphtho[2,1-b]furan-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

044199-500MG 044199-1G 044199-5G
101864-678EA 375.38 USD
101864-678 101864-680 101864-682
1,2-Dihydronaphtho[2,1-b]furan-2-carbohydrazide ≥95%
1,2-Dihydronaphtho[2,1-b]furan-2-carbohydrazide

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₂
Melting Pt: 183…185 °C
MDL Number: MFCD03001255
CAS Number: 400878-27-5

Order Now

Specification Test Results

Melting point 183-185°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR