6-[(2-Methoxyanilino)carbonyl]-3-cyclohexene-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

044196-500MG 044196-1G 044196-5G
101864-660EA 181.54 USD
101864-660 101864-662 101864-664
6-[(2-Methoxyanilino)carbonyl]-3-cyclohexene-1-carboxylic acid ≥95%
6-[(2-Methoxyanilino)carbonyl]-3-cyclohexene-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₇NO₄
Melting Pt: 143…144 °C
MDL Number: MFCD01062916
CAS Number: 326920-49-4

Order Now

Specification Test Results

Melting point 143-144°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR