2-(4-Pyridinyl)-4-pyrimidinamine ≥95%

Supplier: Matrix Scientific

044194-500MG 044194-1G 044194-5G
101864-648EA 375.38 USD
101864-648 101864-650 101864-652
2-(4-Pyridinyl)-4-pyrimidinamine ≥95%
2-(4-Pyridinyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₉H₈N₄
Melting Pt: 262…264 °C
MDL Number: MFCD00119822
CAS Number: 61310-29-0

Order Now

Specification Test Results

Melting point 262-264°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR