1-{4-[(2,4-Dichlorobenzyl)oxy]phenyl}-1-ethanol ≥95%

Supplier: Matrix Scientific

044193-500MG 044193-1G 044193-5G
101864-642EA 375.38 USD
101864-642 101864-644 101864-646
1-{4-[(2,4-Dichlorobenzyl)oxy]phenyl}-1-ethanol ≥95%
1-{4-[(2,4-Dichlorobenzyl)oxy]phenyl}-1-ethanol

Hazard: Irritant

Formula: C₁₅H₁₄Cl₂O₂
Melting Pt: 62…64 °C
MDL Number: MFCD03001253
CAS Number: 61292-28-2

Order Now

Specification Test Results

Melting point 62-64°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR