5-Amino-3-methylisoxazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

044192-500MG 044192-1G 044192-5G
101864-636EA 66.15 USD
101864-636 101864-638 101864-640
5-Amino-3-methylisoxazole-4-carbonitrile ≥95%
5-Amino-3-methylisoxazole-4-carbonitrile

Hazard: Irritant

Formula: C₅H₅N₃O
Melting Pt: 227…230 °C
MDL Number: MFCD00027378
CAS Number: 35261-01-9

Order Now

Specification Test Results

Melting point 227-230°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR