1-Methyl-2-(4-methylpiperazino)-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044191-500MG 044191-1G 044191-5G
101864-630EA 556.92 USD
101864-630 101864-632 101864-634
1-Methyl-2-(4-methylpiperazino)-1H-indole-3-carbaldehyde ≥95%
1-Methyl-2-(4-methylpiperazino)-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₅H₁₉N₃O
MW: 257.34 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD05663789
CAS Number: 886360-89-0

Order Now

Specification Test Results

Melting point 143-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR