Methyl-3-{[(4-methylphenyl)sulfonyl]oxy}-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044190-500MG 044190-1G
101864-626EA 375.38 USD
101864-626 101864-628
Methyl-3-{[(4-methylphenyl)sulfonyl]oxy}-2-thiophenecarboxylate ≥95%
Methyl-3-{[(4-methylphenyl)sulfonyl]oxy}-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₂O₅S₂
Melting Pt: 103…105 °C
MDL Number: MFCD00178121
CAS Number: 181226-89-1

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR