Methyl-2,6-dihydroxy-4-methylbenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,6-dihydroxy-4-methylbenzoate

044188-500MG 044188-1G 044188-5G
101864-614EA 278.46 USD
101864-614 101864-616 101864-618
Methyl-2,6-dihydroxy-4-methylbenzenecarboxylate ≥95%
Methyl-2,6-dihydroxy-4-methylbenzenecarboxylate

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 98…99 °C
MDL Number: MFCD00012063
CAS Number: 16846-10-9

Order Now

Specification Test Results

Melting point 98-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR