2,6-Dihydroxy-4-methylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dihydroxy-p-toluic Acid

044187-500MG 044187-1G 044187-5G
101864-608EA 181.54 USD
101864-608 101864-610 101864-612
2,6-Dihydroxy-4-methylbenzoic acid ≥95%
2,6-Dihydroxy-4-methylbenzoic acid

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 165…166 °C
MDL Number: MFCD00184686
CAS Number: 480-67-1

Order Now

Specification Test Results

Melting point 165-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR