Methyl-4-(4-chlorophenyl)-2,4-dioxobutanoate ≥95%

Supplier: Matrix Scientific

044186-500MG 044186-1G 044186-5G
101864-602EA 141.54 USD
101864-602 101864-604 101864-606
Methyl-4-(4-chlorophenyl)-2,4-dioxobutanoate ≥95%
Methyl-4-(4-chlorophenyl)-2,4-dioxobutanoate

Hazard: Irritant

Formula: C₁₁H₉ClO₄
Melting Pt: 118…120 °C
MDL Number: MFCD03001315
CAS Number: 39757-35-2

Order Now

Specification Test Results

Melting point 118-120°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR