Methyl-2-(4-chloro-2-methylphenoxy)-3-methoxyacrylate ≥95%

Supplier: Matrix Scientific

044184-500MG 044184-1G 044184-5G
101864-596EA 556.92 USD
101864-596 101864-598 101864-600
Methyl-2-(4-chloro-2-methylphenoxy)-3-methoxyacrylate ≥95%
Methyl-2-(4-chloro-2-methylphenoxy)-3-methoxyacrylate

Hazard: Irritant

Formula: C₁₂H₁₃ClO₄
Melting Pt: 68 °C
MDL Number: MFCD08443943
CAS Number: 255908-42-0

Order Now

Specification Test Results

Melting point 68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR