2-Chloro-4,8-dimethylquinoline ≥95%

Supplier: Matrix Scientific

044182-500MG 044182-1G 044182-5G
101864-586EA 87.69 USD
101864-586 101864-588 101864-590
2-Chloro-4,8-dimethylquinoline ≥95%
2-Chloro-4,8-dimethylquinoline

Hazard: Irritant

Formula: C₁₁H₁₀ClN
MW: 191.66 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD00760267
CAS Number: 3913-17-5

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR