4,8-Dimethyl-2-quinolinol ≥95%

Supplier: Matrix Scientific

044181-500MG 044181-1G 044181-5G
101864-580EA 200 USD
101864-580 101864-582 101864-584
4,8-Dimethyl-2-quinolinol ≥95%
4,8-Dimethyl-2-quinolinol

Hazard: Irritant

Formula: C₁₁H₁₁NO
Melting Pt: 219-221°C
MDL Number: MFCD00085676
CAS Number: 5349-78-0

Order Now

Specification Test Results

Melting point 219-221°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR