1-Methyl-2-(2,4,5-trichlorophenoxy)-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044179-500MG 044179-1G 044179-5G
101864-570EA 747.69 USD
101864-570 101864-572 101864-574
1-Methyl-2-(2,4,5-trichlorophenoxy)-1H-indole-3-carbaldehyde ≥95%
1-Methyl-2-(2,4,5-trichlorophenoxy)-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₀Cl₃NO₂
MW: 354.62 g/mol
Melting Pt: 160…163 °C
MDL Number: MFCD01315836
CAS Number: 338416-88-9

Order Now

Specification Test Results

Melting point 160-163°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR